Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF3i Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106410
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106410 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106410 Supplier Invitrogen™ Supplier No. PA5106410
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total EIF3I.

Component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis. The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S preinitiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of post termination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF3i
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Eif3i
Gene Accession No. B0BNA7, Q13347, Q9QZD9
Gene Alias 36kDa; D4Ertd632e; eIF3 p36; eIF-3-beta; eIF3-beta; Eif3i; eIF3-p36; Eif3s2; Eukaryotic translation initiation factor 3 subunit 2; eukaryotic translation initiation factor 3 subunit I; eukaryotic translation initiation factor 3, subunit 1; eukaryotic translation initiation factor 3, subunit 2 (beta); eukaryotic translation initiation factor 3, subunit 2 (beta, 36kD); eukaryotic translation initiation factor 3, subunit 2 beta, 36kDa; eukaryotic translation initiation factor 3, subunit I; predicted protein of HQ2242; PRO2242; subuni; TGF-beta receptor binding protein; TGF-beta receptor-interacting protein 1; TGFbeta receptor-interacting protein 1; TRIP1; TRIP-1
Gene Symbols Eif3i
Host Species Rabbit
Immunogen A synthesized peptide derived from human EIF3I(Accession Q13347), corresponding to amino acid residues A273-F323.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54709, 682390, 8668
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.