Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF3J Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$149.33 - $394.26

Specifications

Antigen EIF3J
Concentration 0.5 mg/mL
Dilution Western Blot 0.5-1.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224572SS
SDP
View Documents
Novus Biologicals
NBP224572SS
0.025 mg
Each for $149.33
Only null left
Add to Cart
 
NBP224572
SDP
View Documents
Novus Biologicals
NBP224572
0.1 mg
Each for $394.26
Only null left
Add to Cart
 
Description

Description

EIF3J Polyclonal specifically detects EIF3J in Human, Mouse, Canine, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

EIF3J
Western Blot 0.5-1.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Polyclonal
Rabbit
Signal Transduction
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
eIF3 p35, eIF3-alpha, eIF3j, eIF3-p35, EIF3S1eIF-3-alpha, Eukaryotic translation initiation factor 3 subunit 1, eukaryotic translation initiation factor 3 subunit J, eukaryotic translation initiation factor 3, subunit 1 (alpha, 35kD), eukaryotic translation initiation factor 3, subunit 1 alpha, 35kDa, eukaryotic translation initiation factor 3, subunit J
EIF3J
IgG
Affinity Purified
The GPR19 amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_b), 95% in chimpanzee, 90% in monkey, 76% in mouse and 73% homologous in horse.
0.5 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse, Canine, Equine, Primate
O75822
8669
A portion of amino acids 230-258 of human eIF3J was used as the immunogen for this antibody.
Primary
Store at -20C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.