Learn More
eIF3J/EIF3S1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | eIF3J/EIF3S1 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2412
![]() |
Bethyl Laboratories, Inc
IHC00617T |
10 μL |
Each for $151.31
|
|
|||||
50-157-2411
![]() |
Bethyl Laboratories, Inc
IHC00617 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Dog, Horse, Guinea pig_10141, Pig, Panda, Orangutan, Monkey, Gorilla, Naked mole rat
Eukaryotic initiation factor-3 (EIF3) has a molecular mass of about 600 kD and contains 13 nonidentical protein subunits, including EIF3J. EIF3 plays a central role in binding of initiator methionyl-tRNA and mRNA to the 40S ribosomal subunit to form the 40S initiation complex (Fraser et al., 2004 [PubMed 14688252]; Fraser et al., 2007 [PubMed 17588516]).Specifications
eIF3J/EIF3S1 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
eIF-3-alpha; eIF3 p35; eukaryotic translation initiation factor 3 subunit 1; eukaryotic translation initiation factor 3 subunit J; eukaryotic translation initiation factor 3, subunit 1 (alpha, 35kD); eukaryotic translation initiation factor 3, subunit 1 alpha, 35kDa | |
EIF3J, Eif3j1, Eif3j2 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
O75822, Q3UGC7, Q66JS6 | |
100042807, 78655, 8669 | |
Between 208 and 258 | |
Primary | |
4° C | |
EIF3J |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.