Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF3j Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597138
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597138 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597138 Supplier Invitrogen™ Supplier No. PA597138
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: SKOV3, HeLa, Jurkat, A-549, U-87MG, LO2, Mouse testis, Mouse liver; Cellular Location: Cytoplasm.

Eukaryotic initiation factor-3 (EIF3) has a molecular mass of about 600 kD and contains 13 nonidentical protein subunits, including EIF3J. EIF3 plays a central role in binding of initiator methionyl-tRNA and mRNA to the 40S ribosomal subunit to form the 40S initiation complex (Fraser et al., 2004; Fraser et al., 2007).
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF3j
Applications ELISA, Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene EIF3J
Gene Accession No. A0JPM9, O75822, Q3UGC7, Q66JS6
Gene Alias 1810016I04Rik; 2700079K05Rik; AA409117; AA409446; C78575; eIF3 p35; eIF3 p35 {ECO:0000255; eIF-3-alpha; eIF3-alpha; eIF-3-alpha {ECO:0000255; eIF-3-alpha-A; eIF-3-alpha-B; Eif3j; eIF3j {ECO:0000255; Eif3j1; Eif3j-1; Eif3j2; Eif3j-2; eIF3j-A; eIF3j-B; eIF3-p35; EIF3S1; Eif3s1-1; Eif3s1-2; ENSMUSG00000043424; Eukaryotic translation initiation factor 3 subunit 1; eukaryotic translation initiation factor 3 subunit 1 {ECO:0000255; eukaryotic translation initiation factor 3 subunit 1-A; Eukaryotic translation initiation factor 3 subunit 1-B; eukaryotic translation initiation factor 3 subunit J; eukaryotic translation initiation factor 3 subunit J {ECO:0000255; Eukaryotic translation initiation factor 3 subunit J-A; Eukaryotic translation initiation factor 3 subunit J-B; eukaryotic translation initiation factor 3, subunit 1 (alpha, 35kD); eukaryotic translation initiation factor 3, subunit 1 alpha; eukaryotic translation initiation factor 3, subunit 1 alpha, 35kDa; eukaryotic translation initiation factor 3, subunit J; eukaryotic translation initiation factor 3, subunit J1; eukaryotic translation initiation factor 3, subunit J2; Gm9781; HAMAP-Rule:MF_03009}; PRO0391
Gene Symbols EIF3J, Eif3j1, Eif3j2
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100042807, 691947, 78655, 8669
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.