Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eIF3K Antibody (01), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306567APC
Description
eIF3K Monoclonal antibody specifically detects eIF3K in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
eIF3K | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunocytochemistry | |
01 | |
Immunocytochemistry/ Immunofluorescence | |
ARG134, eIF3k, EIF3-p28, EIF3S12eIF-3 p28, Eukaryotic translation initiation factor 3 subunit 12, eukaryotic translation initiation factor 3 subunit K, eukaryotic translation initiation factor 3, subunit 12, eukaryotic translation initiation factor 3, subunit K, HSPC029, M9, MSTP001, muscle specific, Muscle-specific gene M9 protein, PLAC24, PLAC-24, PLAC-24eIF-3 p25, PRO1474, PTD001 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human eIF3K (Uniprot#: Q9UBQ5; Ala2-Gln218). | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
27335 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction