Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eIF3K Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 5 publications
Supplier: Novus Biologicals NB10093304
Description
eIF3K Polyclonal specifically detects eIF3K in Human, Chinese Hamster samples. It is validated for Western Blot, Immunoprecipitation.Specifications
eIF3K | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
ARG134, eIF3k, EIF3-p28, EIF3S12eIF-3 p28, Eukaryotic translation initiation factor 3 subunit 12, eukaryotic translation initiation factor 3 subunit K, eukaryotic translation initiation factor 3, subunit 12, eukaryotic translation initiation factor 3, subunit K, HSPC029, M9, MSTP001, muscle specific, Muscle-specific gene M9 protein, PLAC24, PLAC-24, PLAC-24eIF-3 p25, PRO1474, PTD001 | |
Rabbit | |
Affinity Purified | |
RUO | |
27335 | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate | |
Q9UBQ5 | |
EIF3K | |
The immunogen recognized by this antibody maps to a region between residue 168 and 218 of human eukaryotic translation initiation factor 3, subunit K (eukaryotic translation initiation factor 3, subunit 12) using the numbering given in entry NP_037366.1 ( | |
0.1 mL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction