Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF3K/EIF3S12 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501559641
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-9641 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-9641 Supplier Bethyl Laboratories, Inc Supplier No. A301763AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Bovine

Mammalian translation initiation factor 3 (eIF3) is a multi subunit complex containing at least 12 subunits with an apparent aggregate mass of approximately 700 kDa. eIF3k, the smallest subunit of eukaryotic initiation factor 3 (eIF3), interacts with several other subunits of eIF3 and the 40 S ribosomal subunit. eIF3k is conserved among high eukaryotes, including mammals, insects, and plants, and it is ubiquitously expressed in human tissues.

Specifications

Antigen eIF3K/EIF3S12
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene EIF3K
Gene Accession No. Q9UBQ5
Gene Alias ARG134, EIF3-p28, EIF3S12, HSPC029, M9, MSTP001, PLAC-24, PLAC24, PRO1474, PTD001, eIF-3 p25, eIF-3 p28, eukaryotic translation initiation factor 3 subunit 12, eukaryotic translation initiation factor 3 subunit K, eukaryotic translation initiation factor 3, subunit 12, muscle specific, muscle-specific gene M9 protein
Gene Symbols EIF3K
Host Species Rabbit
Immunogen Between 168 and 218
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27335
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.