Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eIF3k Polyclonal Antibody
eIF3k Polyclonal Antibody
Supplier: Thermo Scientific PA598862
Description
eIF3k Polyclonal Antibody for Western Blot, IHC (P), IP, ELISA

Specifications
eIF3k | |
Polyclonal | |
Unconjugated | |
EIF3K | |
1200009C21Rik; ARG134; eIF-3 p25; eIF-3 p28; eIF3k; EIF3-p28; Eif3s12; eukaryotic translation initiation factor 3 subunit 12; Eukaryotic translation initiation factor 3 subunit K; eukaryotic translation initiation factor 3, subunit 12; eukaryotic translation initiation factor 3, subunit K; HSPC029; M9; MSTP001; muscle specific; muscle-specific gene M9 protein; PLAC24; PLAC-24; PRO1474; PTD001 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27335 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9UBQ5 | |
EIF3K | |
Recombinant Human Eukaryotic translation initiation factor 3 subunit K protein (1-218AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction