Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF3k Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118852
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118852 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5118852 Supplier Invitrogen™ Supplier No. PA5118852
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total EIF3K.

The Wilms tumor suppressor gene WT1 appears to play a role in both transcriptional and posttranscriptional regulation of certain cellular genes. This gene encodes a WT1-associating protein, which is a ubiquitously expressed nuclear protein. Like WT1 protein, this protein is localized throughout the nucleoplasm as well as in speckles and partially colocalizes with splicing factors. Alternative splicing of this gene results in three transcript variants, two of which encode the same isoform.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF3k
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EIF3K
Gene Accession No. Q9DBZ5, Q9UBQ5
Gene Alias 1200009C21Rik; ARG134; eIF-3 p25; eIF-3 p28; eIF3k; EIF3-p28; Eif3s12; eukaryotic translation initiation factor 3 subunit 12; Eukaryotic translation initiation factor 3 subunit K; eukaryotic translation initiation factor 3, subunit 12; eukaryotic translation initiation factor 3, subunit K; HSPC029; M9; MSTP001; muscle specific; muscle-specific gene M9 protein; PLAC24; PLAC-24; PRO1474; PTD001
Gene Symbols EIF3K
Host Species Rabbit
Immunogen A synthesized peptide derived from human EIF3K(Accession Q9UBQ5), corresponding to amino acid residues M1-L44.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27335, 292762, 73830
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.