Learn More
Invitrogen™ eIF3k Polyclonal Antibody

Description
The antibody detects endogenous levels of total EIF3K protein.
Specifications
Specifications
| Antigen | eIF3k |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | EIF3K |
| Gene Accession No. | Q9UBQ5 |
| Gene Alias | 1200009C21Rik; ARG134; eIF-3 p25; eIF-3 p28; eIF3k; EIF3-p28; Eif3s12; eukaryotic translation initiation factor 3 subunit 12; Eukaryotic translation initiation factor 3 subunit K; eukaryotic translation initiation factor 3, subunit 12; eukaryotic translation initiation factor 3, subunit K; HSPC029; M9; MSTP001; muscle specific; muscle-specific gene M9 protein; PLAC24; PLAC-24; PRO1474; PTD001 |
| Gene Symbols | EIF3K |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.