Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EIF3M Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100244
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100244 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100244 Supplier Invitrogen™ Supplier No. PA5100244
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total EIF3M.

This gene encodes a protein that is part of the eurkaryotic translation initiation factor 3 complete (eIF-3) required for protein synthesis. Elevated levels of the encoded protein are present in cancer cell lines. Inactivation of the encoded protein has been shown to interfere with translation of herpes virus mRNAs by preventing the association of mRNAs with the ribosomes. A pseudogene of this gene is located on the X chromosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EIF3M
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EIF3M
Gene Accession No. Q7L2H7, Q99JX4
Gene Alias B5; B5 receptor; dendritic cell protein; dendritic cell protein GA17; eIF3m; Eukaryotic translation initiation factor 3 subunit M; eukaryotic translation initiation factor 3, subunit M; fetal lung protein B5; FLJ29030; GA17; HFLB5; hFL-B5; PCI domain containing 1 (herpesvirus entry mediator); PCI domain-containing protein 1; PCID1; PNAS-125; RGD1565840; Tango7; transport and golgi organization 7 homolog
Gene Symbols EIF3M
Host Species Rabbit
Immunogen A synthesized peptide derived from human EIF3M(Accession Q7L2H7), corresponding to amino acid residues T338-T374.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10480, 295975, 98221
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.