Learn More
EIF3M Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | EIF3M |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-8603
![]() |
Bethyl Laboratories, Inc
A304761AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine and Orangutan
HFLB5 encodes a broadly expressed protein containing putative membrane fusion domains that acts as a receptor or coreceptor for entry of herpes simplex virus (HSV) (Perez et al., 2005 [PubMed 15919898]).Specifications
EIF3M | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
EIF3M; B5 receptor; PCID1; PCI domain containing 1 (herpesvirus entry mediator); FLJ29030; GA17; hfl-B5; dendritic cell protein; fetal lung protein B5; eukaryotic translation initiation factor 3, subunit M; B5; PCID1 | |
Between 1 and 50 | |
Primary | |
4° C | |
EIF3M |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q7L2H7, Q99JX4 | |
10480, 98221 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.