Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EIF3S3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | EIF3S3 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
EIF3S3 Polyclonal specifically detects EIF3S3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
EIF3S3 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
eIF-3-gamma, eIF3-gamma, eIF3h, eIF3-p40, EIF3S3eIF3 p40 subunit, Eukaryotic translation initiation factor 3 subunit 3, eukaryotic translation initiation factor 3 subunit H, eukaryotic translation initiation factor 3, subunit 2 (beta, 36kD), eukaryotic translation initiation factor 3, subunit 3 (gamma, 40kD), eukaryotic translation initiation factor 3, subunit 3 gamma, 40kDa, eukaryotic translation initiation factor 3, subunit H, MGC102958 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 80-340 of human EIF3S3 (NP_003747.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Angiogenesis, Cancer | |
PBS with 50% glycerol, pH7.3. | |
8667 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title