Learn More
Eif3s6 Int6 Polyclonal Antibody, Rockland™

Rabbit Polyclonal Antibody
Supplier: Rockland Immunochemicals 600401A62

Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is specific for mouse eIF3S6/Int6 protein. A BLAST analysis was used to suggest cross-reactivity with most eIF3Se/Int6 isoforms from mouse, human, rat, dog, bovine, and monkey based on 100% homology with the immunizing sequence. Cross-reactivity with EIF3S6/Int6 from other sources has not been determined.
Component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis. The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S preinitiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of post termination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation. Required for nonsense-mediated mRNA decay (NMD); may act in conjunction with UPF2 to divert mRNAs from translation to the NMD pathway. May interact with MCM7 and EPAS1 and regulate the proteasome-mediated degradation of these proteins.Specifications
Eif3s6 Int6 | |
Polyclonal | |
Unconjugated | |
EIF3E | |
48kDa; BOS_14000; eIF-3 p48; Eif3e; eIF3-p46; eIF3-p48; Eif3s6; Eukaryotic translation initiation factor 3 subunit 6; Eukaryotic translation initiation factor 3 subunit E; eukaryotic translation initiation factor 3 subunit E isoform 2 transcript; eukaryotic translation initiation factor 3, subunit 6 (48kD); eukaryotic translation initiation factor 3, subunit 6 48kDa; eukaryotic translation initiation factor 3, subunit E; Int6; mammary tumor integration site 6; mammary tumor-associated protein INT6; Mammary tumor-associated protein INT-6; MMTV integration site 6; murine mammary tumor integration site 6 (oncogene homolog); NEWGENE_1311521; Viral integration site protein INT-6; Viral integration site protein INT-6 homolog | |
Rabbit | |
RUO | |
103692906, 16341, 3646, 475070, 534165 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.8 mg/mL | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
P60228, P60229, Q3T102, Q641X8 | |
EIF3E | |
100 μg | |
Primary | |
Bovine, Canine, Human, Monkey, Mouse, Rat | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.