Learn More
Invitrogen™ EIF4A1/EIF4A2/EIF4A3 Polyclonal Antibody

Description
Positive Samples: 293T, HeLa, HepG2, MCF7, C2C12, Mouse brain, Rat brain Immunogen sequence: MSASQDSRSR DNGPDGMEPE GVIESNWNEI VDSFDDMNLS ESLLRGIYAY GFEKPSAIQQ RAILPCIKGY DVIAQAQSGT GKTATFAISI LQQIELDLKA.
Specifications
Specifications
| Antigen | EIF4A1/EIF4A2/EIF4A3 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.638 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 |
| Gene | EIF4A3 |
| Gene Accession No. | P10630, P38919, P60842, P60843, Q14240, Q3B8Q2, Q5RKI1, Q91VC3 |
| Gene Alias | 2400003O03Rik; 4833432N07Rik; ATP-dependent RNA helicase DDX48; ATP-dependent RNA helicase eIF4A-1; ATP-dependent RNA helicase eIF4A-2; ATP-dependent RNA helicase eIF4A-3; BM-010; Ddx2a; Ddx2b; Ddx48; DEAD (Asp-Glu-Ala-Asp) box polypeptide 48; DEAD box protein 48; Eif4; EIF4A; EIF-4A; EIF4A1; EIF4A2; Eif4a3; eIF-4A-I; eIF4A-I; eIF-4A-II; eIF4A-II; eIF4AIII; eIF-4A-III; eIF4A-III; EIF4F; eukaryotic initiation factor 4AI; Eukaryotic initiation factor 4A-I; eukaryotic initiation factor 4A-II; Eukaryotic initiation factor 4A-III; Eukaryotic initiation factor 4A-III, N-terminally processed; Eukaryotic initiation factor 4A-like NUK-34; Eukaryotic translation initiation factor 4A isoform 3; eukaryotic translation initiation factor 4A, isoform 1; eukaryotic translation initiation factor 4A, isoform 2; eukaryotic translation initiation factor 4A, isoform 3; eukaryotic translation initiation factor 4A1; eukaryotic translation initiation factor 4A2; eukaryotic translation initiation factor 4A3; hNMP 265; initiation factor eIF-4A long form; KIAA0111; mKIAA0111; MUK34; NMP 265; NMP265; Nuclear matrix protein 265; NUK34; protein synthesis initiation factor 4AII; RCPS |
| Gene Symbols | EIF4A1, EIF4A2, EIF4A3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.