Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EIF4A1/EIF4A2/EIF4A3 Recombinant Rabbit Monoclonal Antibody (10P2J5)
GREENER_CHOICE

Catalog No. MA569275
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA569275 20 μL
FEMA551538 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA569275 Supplier Invitrogen™ Supplier No. MA569275
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: MSASQDSRSR DNGPDGMEPE GVIESNWNEI VDSFDDMNLS ESLLRGIYAY GFEKPSAIQQ RAILPCIKGY DVIAQAQSGT GKTATFAISI LQQIELDLKA.

The eIF4A proteins are involved in many aspects of RNA metabolism due to their RNA-binding capacity and ATPase activity. eIF4A1 and eIF4A2 are cytoplasmic, whereas eIF4A3 is nuclear-localized. In the current model of translation initiation, eIF4A unwinds RNA secondary structures in the 5'-UTR of mRNAs which is necessary to allow efficient binding of the small ribosomal subunit, and subsequent scanning for the initiator codon.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EIF4A1/EIF4A2/EIF4A3
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 10P2J5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Gene EIF4A3
Gene Accession No. P10630, P38919, P60842, P60843, Q14240, Q3B8Q2, Q5RKI1, Q91VC3
Gene Alias 2400003O03Rik; 4833432N07Rik; ATP-dependent RNA helicase DDX48; ATP-dependent RNA helicase eIF4A-1; ATP-dependent RNA helicase eIF4A-2; ATP-dependent RNA helicase eIF4A-3; BM-010; Ddx2a; Ddx2b; Ddx48; DEAD (Asp-Glu-Ala-Asp) box polypeptide 48; DEAD box protein 48; Eif4; EIF4A; EIF-4A; EIF4A1; EIF4A2; Eif4a3; eIF-4A-I; eIF4A-I; eIF-4A-II; eIF4A-II; eIF4AIII; eIF-4A-III; eIF4A-III; EIF4F; eukaryotic initiation factor 4AI; Eukaryotic initiation factor 4A-I; eukaryotic initiation factor 4A-II; Eukaryotic initiation factor 4A-III; Eukaryotic initiation factor 4A-III, N-terminally processed; Eukaryotic initiation factor 4A-like NUK-34; Eukaryotic translation initiation factor 4A isoform 3; eukaryotic translation initiation factor 4A, isoform 1; eukaryotic translation initiation factor 4A, isoform 2; eukaryotic translation initiation factor 4A, isoform 3; eukaryotic translation initiation factor 4A1; eukaryotic translation initiation factor 4A2; eukaryotic translation initiation factor 4A3; hNMP 265; initiation factor eIF-4A long form; KIAA0111; mKIAA0111; MUK34; NMP 265; NMP265; Nuclear matrix protein 265; NUK34; protein synthesis initiation factor 4AII; RCPS
Gene Symbols EIF4A1, EIF4A2, EIF4A3
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13681, 13682, 192170, 1973, 1974, 287436, 303831, 688288, 9775
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.