Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF4A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530216
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530216 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530216 Supplier Invitrogen™ Supplier No. PA530216
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (95%), Dog (100%), Pig (100%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

eIF4A2 is a 407 amino acid ATP-dependent RNA helicase belonging to the DEAD box helicase family. It contains one helicase ATP-binding domain and a helicase C-terminal domain. eIF4A2, a subunit of the eIF4F complex, acts as a protein-synthesis initiation factor and thus plays an important role in the binding of mRNA to the 43S pre-initiation complex of ribosomes. Human eIF4A2 interacts with the NS5B protein of Hepatitis C virus (HCV) and facilitates the genomic RNA synthesis of NS5B protein by unwinding the secondary structure of the HCV genome. It is ubiquitously expressed in most of normal tissues, but in variable amounts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF4A1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene EIF4A1
Gene Accession No. P60842, P60843
Gene Alias ATP-dependent RNA helicase eIF4A-1; BM-010; DDX2A; Eif4; EIF4A; EIF-4A; Eif4a1; eIF-4A-I; eIF4A-I; eukaryotic initiation factor 4AI; Eukaryotic initiation factor 4A-I; eukaryotic translation initiation factor 4A, isoform 1; eukaryotic translation initiation factor 4A1; initiation factor eIF-4A long form; QtsA-12280; unnamed protein product
Gene Symbols EIF4A1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human EIF4A1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13681, 1973, 287436
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.