Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF4A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523264
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523264 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523264 Supplier Invitrogen™ Supplier No. PA523264
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

eIF4A2 Polyclonal Antibody for Western Blot, IHC (P)

eIF4A2 is a 407 amino acid ATP-dependent RNA helicase belonging to the DEAD box helicase family. It contains one helicase ATP-binding domain and a helicase C-terminal domain. eIF4A2, a subunit of the eIF4F complex, acts as a protein-synthesis initiation factor and thus plays an important role in the binding of mRNA to the 43S pre-initiation complex of ribosomes. Human eIF4A2 interacts with the NS5B protein of Hepatitis C virus (HCV) and facilitates the genomic RNA synthesis of NS5B protein by unwinding the secondary structure of the HCV genome. It is ubiquitously expressed in most of normal tissues, but in variable amounts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF4A2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA and 0.05% sodium azide
Gene EIF4A2
Gene Accession No. P10630, Q14240, Q8JFP1
Gene Alias 4833432N07Rik; ATP dependent RNA helicase eIF4A 2; ATP-dependent RNA helicase eIF4A-2; BM-010; DDX2B; Eif4; EIF4A; EIF4A1; EIF4A2; EIF4AII; eIF-4A-II; eIF4A-II; EIF4F; Eukaryotic initiation factor 4A-II; eukaryotic translation initiation factor 4A; eukaryotic translation initiation factor 4A isoform 2; eukaryotic translation initiation factor 4A, isoform 2; eukaryotic translation initiation factor 4A1; eukaryotic translation initiation factor 4A2; hypothetical protein; N-terminally processed; protein synthesis initiation factor 4AII; RCJMB04_14a6; translational eukaryotic inititation factor 4AII
Gene Symbols EIF4A2
Host Species Rabbit
Immunogen A portion of amino acids 50-100 of human eIF4A2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13682, 1974, 395232, 488118
Target Species Human, Mouse, Canine, Monkey, Chicken
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.