Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eIF4A2 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS14549R

Description
eIF4A2 is a 407 amino acid ATP-dependent RNA helicase belonging to the DEAD box helicase family. It contains one helicase ATP-binding domain and a helicase C-terminal domain. eIF4A2, a subunit of the eIF4F complex, acts as a protein-synthesis initiation factor and thus plays an important role in the binding of mRNA to the 43S pre-initiation complex of ribosomes. Human eIF4A2 interacts with the NS5B protein of Hepatitis C virus (HCV) and facilitates the genomic RNA synthesis of NS5B protein by unwinding the secondary structure of the HCV genome. It is ubiquitously expressed in most of normal tissues, but in variable amounts.Specifications
eIF4A2 | |
Polyclonal | |
Unconjugated | |
EIF4A2 | |
4833432N07Rik; ATP-dependent RNA helicase eIF4A-2; BM-010; Ddx2b; Eif4; EIF4A; Eif4a2; EIF4AII; eIF-4A-II; eIF4A-II; EIF4F; Eukaryotic initiation factor 4A-II; eukaryotic translation initiation factor 4A, isoform 2; eukaryotic translation initiation factor 4A2; protein synthesis initiation factor 4AII; RCJMB04_14a6; translational eukaryotic inititation factor 4AII | |
Rabbit | |
Protein A | |
RUO | |
1974 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q14240 | |
EIF4A2 | |
KLH conjugated synthetic peptide derived from human eIF4A2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction