Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EIF4A3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | EIF4A3 |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
EIF4A3 Polyclonal specifically detects EIF4A3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
EIF4A3 | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
ATP-dependent RNA helicase DDX48, ATP-dependent RNA helicase eIF4A-3, DDX48, DEAD (Asp-Glu-Ala-Asp) box polypeptide 48, DEAD box protein 48, DKFZp686O16189, EC 3.6.1, EC 3.6.4.13, eIF4AIII, eIF-4A-III, eIF4A-III, eukaryotic initiation factor 4A-III, Eukaryotic initiation factor 4A-like NUK-34, eukaryotic translation initiation factor 4A, Eukaryotic translation initiation factor 4A isoform 3, eukaryotic translation initiation factor 4A3, hNMP 265, KIAA0111eukaryotic translation initiation factor 4A, isoform 3, MGC10862, NMP 265, NMP265, Nuclear matrix protein 265, NUK34 | |
EIF4A3 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
9775 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:MATTATMATSGSARKRLLKEEDMTKVEFETSEEVDVTPTFDTMGLREDLLRGIYAYGFEKPSAIQQRAIKQIIK | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title