Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF4A3 Mouse anti-Human, Mouse, Rat, Clone: 1G6H9, ProteinTech
SDP

Catalog No. 502239796 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-223-9796 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-223-9796 Supplier Proteintech Group Inc Supplier No. 677401IG150UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Aliquoting is unnecessary for -20°C storage.

EIF4A3 Monoclonal antibody specifically detects EIF4A3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen EIF4A3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1G6H9
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene EIF4A3
Gene Accession No. P38919, Q3B8Q2, Q91VC3
Gene Alias 2400003O03Rik; ATP-dependent RNA helicase DDX48; ATP-dependent RNA helicase eIF4A-3; Ddx48; DEAD (Asp-Glu-Ala-Asp) box polypeptide 48; DEAD box protein 48; Eif4a3; eIF4AIII; eIF-4A-III; eIF4A-III; eukaryotic initiation factor 4AIII; Eukaryotic initiation factor 4A-III; Eukaryotic initiation factor 4A-III, N-terminally processed; Eukaryotic initiation factor 4A-III-like protein; eukaryotic initiation factor 4A-like NUK-34; eukaryotic translation initiation factor 4A; Eukaryotic translation initiation factor 4A isoform 3; eukaryotic translation initiation factor 4A, isoform 3; eukaryotic translation initiation factor 4A3; hNMP 265; KIAA0111; mKIAA0111; MUK34; NMP 265; NMP265; Nuclear matrix protein 265; NUK34; RCPS; similar to DEAD (Asp-Glu-Ala-Asp) box polypeptide 48; zgc:56139
Gene Symbols EIF4A3
Host Species Mouse
Immunogen EIF4A3 Fusion Protein Ag11130
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192170, 688288, 9775
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.