Learn More
eIF4AIII/EIF4A3 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302981AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Rat, Zebrafish, X. laevis, X. tropicalis, Chicken, Bovine and Pig
eIF4A3 is a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. eIF4A3 is a nuclear matrix protein. Its amino acid sequence is highly similar to the amino acid sequences of the translation initiation factors eIF4AI and eIF4AII, two other members of the DEAD box protein family.Specifications
| eIF4AIII/EIF4A3 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| P38919, Q91VC3 | |
| EIF4A3 | |
| Between 125 and 175 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| EIF4A3 | |
| DDX48, NMP265, NUK34, eIF4AIII, ATP-dependent RNA helicase DDX48, ATP-dependent RNA helicase eIF4A-3, DEAD (Asp-Glu-Ala-Asp) box polypeptide 48, DEAD box protein 48, NMP 265, eIF-4A-III, eIF4A-III, eukaryotic initiation factor 4A-III, eukaryotic initiation factor 4A-like NUK-34, eukaryotic trans | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 192170, 9775 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.