Learn More
Invitrogen™ eIF4B Polyclonal Antibody

Description
Recommended positive controls: NT2D1, IMR32, MCF-7, U87-MG, SK-N-SH, SK-N-AS, 293T, A431, HeLa, HepG2, human EIF4B-transfected 293T. Predicted reactivity: Mouse (97%), Rat (97%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | eIF4B |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.16 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | Eif4b |
| Gene Accession No. | P23588, Q8BGD9 |
| Gene Alias | 2310046H11Rik; AL024095; C85189; Eif4a2; EIF4B; eIF-4B; Eukaryotic translation initiation factor 4B; hCG_2014609; hypothetical protein; PRO1843 |
| Gene Symbols | Eif4b |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.