Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ eIF4E Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584061
Description
Immunogen sequence: MATVEPETTP TPNPPTTEEE KTESNQEVAN PEHYIKHP.
eIF4E is involved in several cellular processes including enhanced translational efficiency, splicing, mRNA stability, and RNA nuclear export. All eukaryotic cellular mRNAs are blocked at their 5-prime ends with the 7-methylguanosine cap structure, m7GpppX (where X is any nucleotide). EIF4E is a eukaryotic translation initiation factor involved in directing ribosomes to the cap structure of mRNAs. It is a 24-kD polypeptide that exists as both a free form and as part of a multiprotein complex termed EIF4F. The EIF4E polypeptide is the rate-limiting component of the eukaryotic translation apparatus and is involved in the mRNA-ribosome binding step of eukaryotic protein synthesis. The other subunits of EIF4F are a 50-kD polypeptide, termed EIF4A, that possesses ATPase and RNA helicase activities, and a 220-kD polypeptide, EIF4G.
Specifications
| eIF4E | |
| Polyclonal | |
| Unconjugated | |
| Eif4e | |
| AUTS19; CBP; EG668879; EIF4E; EIF-4E; EIF4E1; EIF4EL1; Eif4e-ps; EIF4F; eIF-4F 25 kDa subunit; eukaryotic translation initiation factor 4E; eukaryotic translation initiation factor 4E-like 1; eukaryotic translation initiation factor 4E-like protein; eukaryotic translation initiation factor 4-like protein; If4e; MGC111573; mRNA cap-binding protein; OTTHUMP00000219703; OTTHUMP00000219704; OTTHUMP00000219705; translation initiation factor eIF-4E | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1977 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry, Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P06730 | |
| Eif4e | |
| Recombinant protein corresponding to Human EIF4E. Recombinant protein control fragment (Product #RP-104920). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction