Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF4E Rabbit anti-Human, Mouse, Unconjugated, Clone: DBD-5, Abnova™

Catalog No. 89336911 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-336-911 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-336-911 Supplier Abnova Corporation Supplier No. MAB20511
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

EIF4E Monoclonal antibody specifically detects EIF4E in Human, Mouse samples. It is validated for IP, ICC, WB-Ce, IHC-P, IF.

Specifications

Antigen EIF4E
Applications Immunoprecipitation, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone DBD-5
Conjugate Unconjugated
Description eukaryotic translation initiation factor 4E
Gene EIF4E
Gene Alias CBP, EIF4E1, EIF4EL1, EIF4F, MGC111573
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding S209 f human EIF4E
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 1977
Target Species Human, Mouse
Content And Storage Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.