Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EIF4E1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560908
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560908 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560908 Supplier Invitrogen™ Supplier No. PA560908
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EKEEEAAERT PTGEKSPNSP RTLLSLR Highest antigen sequence identity to the following orthologs: Mouse - 44%, Rat - 45%.

Recognizes and binds the 7-methylguanosine-containing mRNA cap during an early step in the initiation of protein synthesis and facilitates ribosome binding by inducing the unwinding of the mRNAs secondary structure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EIF4E1B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EIF4E1B
Gene Accession No. A6NMX2
Gene Alias EIF4E1B; eukaryotic translation initiation factor 4E family member 1B; eukaryotic translation initiation factor 4E type 1B
Gene Symbols EIF4E1B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EIF4E1B. Recombinant protein control fragment (Product #RP-100560).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253314
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.