Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF4E2 Monoclonal Antibody (225CT3.1.3)
GREENER_CHOICE

Catalog No. PIMA537509
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537509 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537509 Supplier Invitrogen™ Supplier No. MA537509
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

eIF4E2 Monoclonal Antibody for Western Blot

Recognizes and binds the 7-methylguanosine-containing mRNA cap during an early step in the initiation (PubMed:9582349, PubMed:17368478, PubMed:25624349). Acts as a repressor of translation initiation (PubMed:22751931). In contrast to EIF4E, it is unable to bind eIF4G (EIF4G1, EIF4G2 or EIF4G3), suggesting that it acts by competing with EIF4E and block assembly of eIF4F at the cap. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF4E2
Applications Western Blot
Classification Monoclonal
Clone 225CT3.1.3
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene EIF4E2
Gene Accession No. O60573
Gene Alias 2700069E09Rik; 4EHP; 4E-LP; AI036339; AV129531; D0H0S6743E; EGK_04904; eIF4E type 2; eIF-4E type 2; Eif4e2; eIF4E-2; EIF4EL3; eIF4E-like cap-binding protein; eIF4E-like protein 4E-LP; eukar; eukaryotic translation initiation factor 4E family member 2; eukaryotic translation initiation factor 4E homologous protein; eukaryotic translation initiation factor 4E like 3; eukaryotic translation initiation factor 4E member 2; eukaryotic translation initiation factor 4E type 2; Eukaryotic translation initiation factor 4E-like 3; h4EHP; IF4e; mRNA cap-binding protein 4EHP; mRNA cap-binding protein type 2; mRNA cap-binding protein type 3
Gene Symbols EIF4E2
Host Species Mouse
Immunogen EIF4E2 recombinant protein.
Purification Method Protein G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9470
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.