Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF4E3 Antibody, ProSci
SDP

Catalog No. 16832006
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-832-006 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-832-006 Supplier ProSci LLC Supplier No. 295170100
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EIF4E3 Polyclonal Antibody specifically detects EIF4E3 in Human, Mouse, Rat samples. It is validated for ELISA, Western Blot

Specifications

Antigen EIF4E3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution EIF4E3 antibody can be used for detection of EIF4E3 by ELISA at 1:62500. EIF4E3 antibody can be used for detection of EIF4E3 by western blot at 0.5 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Formulation Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
Gene Accession No. NP_001128123
Gene Alias EIF4E3, MGC39820, MGC86971, eIF-4E3, eIF4E-3
Host Species Rabbit
Immunogen Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human EIF4E3.
Purification Method Antibody is purified by peptide affinity chromatography method.
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 317649
Target Species Human, Mouse, Rat
Content And Storage For short periods of storage (days) store at 4°C. For longer periods of storage, store EIF4E3 antibody at -20°C. As with any antibody avoid repeat freeze-thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Research Category Other
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.