Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF4E3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103195
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103195 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103195 Supplier Invitrogen™ Supplier No. PA5103195
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total EIF4E3.

EIF4E3 belongs to the EIF4E family of translational initiation factors that interact with the 5-prime cap structure of mRNA and recruit mRNA to the ribosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF4E3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Eif4e3
Gene Accession No. Q8N5X7, Q9DBB5
Gene Alias 1300018P11Rik; AI451927; eIF4E type 3; eIF-4E type 3; Eif4e3; eIF-4E3; eIF4E-3; eukaryotic translation initiation factor 4E class III; eukaryotic translation initiation factor 4E family member 3; eukaryotic translation initiation factor 4E member 3; eukaryotic translation initiation factor 4E type 3; eukaryotic translation initiation factor-3; wu:fc31f11; wu:fd15f11; zgc:92189; zgc:92189 protein
Gene Symbols Eif4e3
Host Species Rabbit
Immunogen A synthesized peptide derived from human EIF4E3(Accession Q8N5X7), corresponding to amino acid residues V88-V138.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 297481, 317649, 66892
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.