Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF4E3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550954
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550954 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550954 Supplier Invitrogen™ Supplier No. PA550954
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EIF4E3 Polyclonal Antibody detects endogenous levels of EIF4E3 protein.

EIF4E3 belongs to the EIF4E family of translational initiation factors that interact with the 5-prime cap structure of mRNA and recruit mRNA to the ribosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF4E3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Eif4e3
Gene Accession No. Q8N5X7, Q9DBB5
Gene Alias 1300018P11Rik; AI451927; eIF4E type 3; eIF-4E type 3; Eif4e3; eIF-4E3; eIF4E-3; eukaryotic translation initiation factor 4E class III; eukaryotic translation initiation factor 4E family member 3; eukaryotic translation initiation factor 4E member 3; eukaryotic translation initiation factor 4E type 3; eukaryotic translation initiation factor-3; wu:fc31f11; wu:fd15f11; zgc:92189; zgc:92189 protein
Gene Symbols Eif4e3
Host Species Rabbit
Immunogen Synthesized peptide derived from the Internal region of human eIF4E3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 297481, 317649, 66892
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.