Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EIF4E3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 172821AP150UL

Description
EIF4E3 belongs to the EIF4E family of translational initiation factors that interact with the 5-prime cap structure of mRNA and recruit mRNA to the ribosome.[supplied by OMIM, Mar 2008].Specifications
EIF4E3 | |
Polyclonal | |
Unconjugated | |
EIF4E3 | |
eIF 4E type 3, eIF 4E3, eIF4E 3, eIF4E type 3, EIF4E3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
297481, 317649, 66892 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8N5X7, Q9DBB5 | |
Eif4e3 | |
EIF4E3 Fusion Protein Ag11056 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction