Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF4EBP1 Rabbit anti-Human, Mouse, Rat, FITC, Clone: A5, Abnova™

Catalog No. 89335604 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-604 10 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-604 Supplier Abnova Corporation Supplier No. MAB18770
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

EIF4EBP1 Monoclonal antibody specifically detects EIF4EBP1 in Human, Mouse, Rat samples. It is validated for Flow Cyt.

Specifications

Antigen EIF4EBP1
Applications Flow Cytometry
Classification Monoclonal
Clone A5
Conjugate FITC
Description eukaryotic translation initiation factor 4E binding protein 1
Gene EIF4EBP1
Gene Alias 4E-BP1, 4EBP1, BP-1, MGC4316, PHAS-I
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding T37/T46 of human EIF4EBP1
Quantity 10 Reactions
Primary or Secondary Primary
Gene ID (Entrez) 1978
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.