Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EIF4EBP3 Monoclonal Antibody (OTI2E7), OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF811912


Description
EIF4EBP3 Monoclonal antibody specifically detects EIF4EBP3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
EIF4EBP3 | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
A2AIV8, Q9EPY0, Q9H257 | |
Mouse | |
Protein A/G | |
RUO | |
332579, 64170, 64171 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Western Blot | |
OTI2E7 | |
Unconjugated | |
CARD9 | |
4EBP3; 4E-BP3; eIF4E-binding protein 3; EIF4EBP3; eukaryotic initiation factor 4E-binding protein 3; eukaryotic translation initiation factor 4E binding protein 3; eukaryotic translation initiation factor 4E-binding protein 3; NG36 | |
Full length human recombinant protein of human CARD9 (NP_434700) produced in HEK293T cell. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction