Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF4ENIF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544757
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544757 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544757 Supplier Invitrogen™ Supplier No. PA544757
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: DSDGLRLLGG RRIGSGRIIS ARTFEKDHRL SDKDLRDLRD RDRERDFKDK Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.

The protein encoded by this gene is a nucleocytoplasmic shuttle protein for the translation initiation factor eIF4E. This shuttle protein interacts with the importin alpha-beta complex to mediate nuclear import of eIF4E. It is predominantly cytoplasmic; its own nuclear import is regulated by a nuclear localization signal and nuclear export signals. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF4ENIF1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene EIF4ENIF1
Gene Accession No. Q9NRA8
Gene Alias 2610509L04Rik; 4E-T; A930019J01Rik; AA410001; AU021239; Clast4; D11Ertd166e; eIF4E transporter; EIF4ENIF1; eIF4E-transporter; Eukaryotic translation initiation factor 4E nuclear import factor 1; eukaryotic translation initiation factor 4E transporter; Eukaryotic translation initiation factor 4E transporter-like protein; FLJ21601; FLJ26551; OTTHUMP00000063276; RGD1560908
Gene Symbols EIF4ENIF1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human EIF4ENIF1 (aa 143-192).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56478
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.