Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF4G Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517422
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517422 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517422 Supplier Invitrogen™ Supplier No. PA517422
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

The protein encoded by this gene is a component of the protein complex EIF4F, which is involved in the recognition of the mRNA cap, ATP-dependent unwinding of 5'-terminal secondary structure, and recruitment of mRNA to the ribosome. Alternative splicing results in five transcript variants encoding four distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF4G
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 7 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene EIF4G1
Gene Accession No. Q04637, Q6NZJ6
Gene Alias DKFZp686A1451; E030015G23Rik; EIF4F; EIF4G; eIF-4G 1; EIF4G1; eIF-4G1; EIF4-gamma; eIF-4-gamma 1; EIF4GI; eIF4GI protein; eucaryotic translation initiation factor 4G; Eukaryotic translation initiation factor 4 gamma 1; eukaryotic translation initiation factor 4 gamma, 1; eukaryotic translation initiation factor 4, gamma 1; OTTHUMP00000210577; OTTHUMP00000210586; OTTHUMP00000210587; OTTHUMP00000210588; OTTHUMP00000210595; OTTHUMP00000210602; OTTHUMP00000210603; P220; PARK18
Gene Symbols EIF4G1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a sequence of human eIF4G.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1981, 208643, 287986
Target Species Human, Mouse, Monkey, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.