Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF4G Polyclonal Antibody, Invitrogen™

Catalog No. PIPA596502
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596502 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596502 Supplier Thermo Scientific Supplier No. PA596502

Rabbit Polyclonal Antibody

Immunogen sequence: PEELLNGAPS PPAVDLSPVS EPEEQAKEVT ASMAPPTIPS ATPATAPSAT SPAQEEEMEE EEEEEEGEAG EAGEAESEKG GEELLPPEST PIPANLSQNL E; Positive Samples: MCF7, U251, BT474, HeLa

The protein encoded by this gene is a component of the protein complex EIF4F, which is involved in the recognition of the mRNA cap, ATP-dependent unwinding of 5'-terminal secondary structure, and recruitment of mRNA to the ribosome. Alternative splicing results in five transcript variants encoding four distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF4G
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.82 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide, pH 7.3
Gene EIF4G1
Gene Accession No. Q04637
Gene Alias DKFZp686A1451, E030015G23Rik, EIF4F, EIF4G, eIF-4G 1, EIF4G1, eIF-4G1, EIF4-gamma, eIF-4-gamma 1, EIF4GI, eIF4GI protein, eucaryotic translation initiation factor 4G, Eukaryotic translation initiation factor 4 gamma 1, eukaryotic translation initiation factor 4 gamma, 1, eukaryotic translation initiation factor 4, gamma 1, OTTHUMP00000210577, OTTHUMP00000210586, OTTHUMP00000210587, OTTHUMP00000210588, OTTHUMP00000210595, OTTHUMP00000210602, OTTHUMP00000210603, P220, PARK18
Gene Symbols EIF4G1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 400-500 of human EIF4G1 (NP_8865533)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1981
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.