Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eIF4G1 Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100268FR
Description
eIF4G1 Polyclonal specifically detects eIF4G1 in Human, Porcine samples. It is validated for Western Blot.Specifications
eIF4G1 | |
Polyclonal | |
Western Blot | |
DKFZp686A1451, EIF4F, eIF-4G 1, EIF-4G1, EIF4-gamma, eIF-4-gamma 1, EIF4GI, EIF4Gp220, eucaryotic translation initiation factor 4G, eukaryotic translation initiation factor 4 gamma 1, eukaryotic translation initiation factor 4 gamma, 1, P220 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Pig | |
IgG |
Western Blot | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
EIF4G1 | |
The immunogen recognized by this antibody maps to a region between residues 575 and 625 of human Eukaryotic Initiation Factor 4 Gamma 1 (NP_886553.2). | |
100 μL | |
Cancer, DNA Repair, DNA replication Transcription Translation and Splicing, mTOR Pathway, Phospho Specific | |
1981 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction