Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eIF4G1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$336.16 - $691.20
Specifications
| Antigen | eIF4G1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
eIF4G1 Polyclonal specifically detects eIF4G1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| eIF4G1 | |
| Polyclonal | |
| Rabbit | |
| Cancer, DNA Repair, DNA replication Transcription Translation and Splicing, mTOR Pathway | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 1981 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:FSALQQAVPTESTDNRRVVQRSSLSRERGEKAGDRGDRLERSERGGDRGDRLDRARTPATKRSFSKEVEERSRERPSQPEGLRKAASLTEDRDRGRDAVKREAA | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| DKFZp686A1451, EIF4F, eIF-4G 1, EIF-4G1, EIF4-gamma, eIF-4-gamma 1, EIF4GI, EIF4Gp220, eucaryotic translation initiation factor 4G, eukaryotic translation initiation factor 4 gamma 1, eukaryotic translation initiation factor 4 gamma, 1, P220 | |
| EIF4G1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title