Learn More
eIF4G1/eIF4GI Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$167.95
Specifications
| Antigen | eIF4G1/eIF4GI |
|---|---|
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-155-9671
|
Bethyl Laboratories, Inc
A301774AT |
20 μL |
Each of 1 for $167.95
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
The protein encoded by this gene is a component of the multi-subunit protein complex EIF4F. This complex facilitates the recruitment of mRNA to the ribosome, which is a rate-limiting step during the initiation phase of protein synthesis. The recognition of the mRNA cap and the ATP-dependent unwinding of 5'-terminal secondary structure is catalyzed by factors in this complex. The subunit encoded by this gene is a large scaffolding protein that contains binding sites for other members of the EIF4F complex. A domain at its N-terminus can also interact with the poly-binding protein, which may mediate the circularization of mRNA during translation. Alternative splicing results in multiple transcript variants, some of which are derived from alternative promoter usage.Specifications
| eIF4G1/eIF4GI | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| DKFZp686A1451, EIF4F, EIF4G, p220 | |
| EIF4G1 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q04637 | |
| 1981 | |
| Between 200 and 250 | |
| Primary | |
| 4° C | |
| EIF4G1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.