Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ eIF4G3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555208
Description
Immunogen sequence: EFDSRRTLTS RGSMGREKND KPLPSATARP NTFMRGGSSK DLLDNQSQEE QRREMLETVK QLTGGVDVER NSTEAERNKT RESAKPEISA MSA Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 78%.
The protein encoded by this gene is thought to be part of the eIF4F protein complex, which is involved in mRNA cap recognition and transport of mRNAs to the ribosome. Interestingly, a microRNA (miR-520c-3p) has been found that negatively regulates synthesis of the encoded protein, and this leads to a global decrease in protein translation and cell proliferation. Therefore, this protein is a key component of the anti-tumor activity of miR-520c-3p.
Specifications
| eIF4G3 | |
| Polyclonal | |
| Unconjugated | |
| EIF4G3 | |
| 1500002J22Rik; 4833436O05; 4930523M17Rik; eIF4G 3; eIF-4G 3; Eif4g3; eIF-4-gamma 3; eIF-4-gamma II; eIF4GII; eukaryotic translation initiation factor 4 gamma 3; eukaryotic translation initiation factor 4 gamma, 3; G1-419-52; repro8; RP11-190H11.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8672 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43432 | |
| EIF4G3 | |
| Recombinant protein corresponding to Human eIF4G3. Recombinant protein control fragment (Product #RP-94018). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction