Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF4G3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555212
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555212 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555212 Supplier Invitrogen™ Supplier No. PA555212
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TEEIMSGGGS RNPTPPIGRP TSTPTPPQQL PSQVPEHSPV VYGTVESAHL AASTPVTAAS DQKQEEKPKP DPVLKSPSPV LRLVLSGEKK EQEGQTSETT AIVS Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.

The protein encoded by this gene is thought to be part of the eIF4F protein complex, which is involved in mRNA cap recognition and transport of mRNAs to the ribosome. Interestingly, a microRNA (miR-520c-3p) has been found that negatively regulates synthesis of the encoded protein, and this leads to a global decrease in protein translation and cell proliferation. Therefore, this protein is a key component of the anti-tumor activity of miR-520c-3p.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF4G3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EIF4G3
Gene Accession No. O43432
Gene Alias 1500002J22Rik; 4833436O05; 4930523M17Rik; eIF4G 3; eIF-4G 3; Eif4g3; eIF-4-gamma 3; eIF-4-gamma II; eIF4GII; eukaryotic translation initiation factor 4 gamma 3; eukaryotic translation initiation factor 4 gamma, 3; G1-419-52; repro8; RP11-190H11.1
Gene Symbols EIF4G3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human eIF4G3. Recombinant protein control fragment (Product #RP-94019).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8672
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.