Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ eIF4G3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531101
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, mouse testis. Predicted reactivity: Human (99%), Mouse (96%), Rat (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is thought to be part of the eIF4F protein complex, which is involved in mRNA cap recognition and transport of mRNAs to the ribosome. Interestingly, a microRNA (miR-520c-3p) has been found that negatively regulates synthesis of the encoded protein, and this leads to a global decrease in protein translation and cell proliferation. Therefore, this protein is a key component of the anti-tumor activity of miR-520c-3p.
Specifications
| eIF4G3 | |
| Polyclonal | |
| Unconjugated | |
| EIF4G3 | |
| 1500002J22Rik; 4833436O05; 4930523M17Rik; eIF4G 3; eIF-4G 3; Eif4g3; eIF-4-gamma 3; eIF-4-gamma II; eIF4GII; eukaryotic translation initiation factor 4 gamma 3; eukaryotic translation initiation factor 4 gamma, 3; G1-419-52; repro8; RP11-190H11.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 230861, 298573, 8672 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.26 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
| O43432, Q80XI3 | |
| EIF4G3 | |
| Recombinant protein encompassing a sequence within the center region of human EIF4G3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction