Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eIF4H Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299470MFV450
Description
eIF4H Polyclonal antibody specifically detects eIF4H in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
eIF4H | |
Polyclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence | |
eukaryotic translation initiation factor 4H, KIAA0038eIF-4H, WBSCR1, Williams-Beuren syndrome chromosome region 1, WSCR1Williams-Beuren syndrome chromosomal region 1 protein | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human eIF4H. | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
7458 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunocytochemistry | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction