Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eIF4H Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP157248
Description
eIF4H Polyclonal specifically detects eIF4H in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
eIF4H | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
eukaryotic translation initiation factor 4H, KIAA0038eIF-4H, WBSCR1, Williams-Beuren syndrome chromosome region 1, WSCR1Williams-Beuren syndrome chromosomal region 1 protein | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Canine: 100%; Human: 100%; Mouse: 92%; Rat: 85%; Trichomonas vaginalis: 84%; Chicken: 84%; Western clawed frog: 81%; Xenopus: 81%; Alistipes sp. HGB5: 76%;. | |
Human, Mouse, Rat, Pig, Canine, Equine | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
Q15056 | |
EIF4H | |
Synthetic peptides corresponding to WBSCR1 The peptide sequence was selected from the middle region of WBSCR1. Peptide sequence DSRDDFNSGFRDDFLGGRGGSRPGDRRTGPPMGSRFRDGPPLRGSNMDFR. | |
100 μL | |
DNA replication Transcription Translation and Splicing | |
7458 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction