Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF4H Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583149
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583149 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583149 Supplier Invitrogen™ Supplier No. PA583149
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KDTDKFKGFC YVEFDEVDSL KEALTYDGAL LGDRSLRVDI AEGRKQDKGG FGFRKGGPDD RGMGSSRESR GGWDSRDDFN SG Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 99%.

This gene encodes one of the translation initiation factors, which functions to stimulate the initiation of protein synthesis at the level of mRNA utilization. This gene is deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.23. Alternative splicing of this gene generates 2 transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF4H
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EIF4H
Gene Accession No. Q15056, Q5XI72, Q9WUK2
Gene Alias AU018978; D5Ertd355e; E430026L18Rik; Ef4h; EIF4H; eIF-4H; Eukaryotic translation initiation factor 4H; KIAA0038; mKIAA0038; WBSCR1; Williams-Beuren syndrome chromosomal region 1 protein; Williams-Beuren syndrome chromosomal region 1 protein homolog; Williams-Beuren syndrome chromosome region 1; Williams-Beuren syndrome chromosome region 1 homolog; WSCR1
Gene Symbols EIF4H
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EIF4H. Recombinant protein control fragment (Product #RP-95110).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22384, 288599, 7458
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.