Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF4H Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501726794 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6794 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6794 Supplier Proteintech Group Inc Supplier No. 110121AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EIF4H has two isoforms. The short isoform (∽25kDa) is the predomit isoform and is expressed alone in liver and skeletal muscle. This is a rabbit anti-EIF4H polyclonal antibody raised against part of N-terminal chain of human EIF4H, and can recognize

This gene encodes one of the translation initiation factors, which functions to stimulate the initiation of protein synthesis at the level of mRNA utilization. This gene is deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.23. Alternative splicing of this gene generates 2 transcript variants.

Specifications

Antigen EIF4H
Applications Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene EIF4H
Gene Accession No. Q15056
Gene Alias eIF 4H, EIF4H, KIAA0038, WBSCR1, WSCR1
Gene Symbols EIF4H
Host Species Rabbit
Immunogen EIF4H Fusion Protein Ag1468
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7458
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.