Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EIF5 Antibody (6D6G6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | EIF5 |
---|---|
Clone | 6D6G6 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, ELISA 1:10000 |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61763
![]() |
Novus Biologicals
NBP261763 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
EIF5 Monoclonal specifically detects EIF5 in Human, Monkey samples. It is validated for Western Blot, ELISA.Specifications
EIF5 | |
1 mg/mL | |
Monoclonal | |
Purified | |
Human, Monkey | |
1983 | |
Purified recombinant fragment of human EIF5 (AA: 1-300) expressed in E. Coli. | |
Primary | |
49.2 kDa |
6D6G6 | |
Western Blot 1:500-1:2000, ELISA 1:10000 | |
Unconjugated | |
Mouse | |
eIF-5, EIF-5A, eukaryotic translation initiation factor 5 | |
EIF5 | |
IgG2a | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title