Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EIF5 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300231MFV610
Description
EIF5 Polyclonal antibody specifically detects EIF5 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
EIF5 | |
Polyclonal | |
ELISA, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
eIF-5, EIF-5A, eukaryotic translation initiation factor 5 | |
Produced in rabbits immunized with purified, recombinant Human EIF5 (Accession#: P55010; Met1-Asp150) | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
1983 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction