Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EIF5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10093312
Description
EIF5 Polyclonal specifically detects EIF5 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation (Negative).Specifications
EIF5 | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
eIF-5, EIF-5A, eukaryotic translation initiation factor 5 | |
Rabbit | |
Affinity Purified | |
RUO | |
1983 | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation | |
NP_892116.2 | |
EIF5 | |
The immunogen recognized by this antibody maps to a region between residue 100 and 150 of human eukaryotic translation initiation factor 5 using the numbering given in entry NP_892116.2 (GeneID 1983). | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction