Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eIF5 Monoclonal Antibody (2F13C4)
GREENER_CHOICE

Catalog No. PIMA549276
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549276 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549276 Supplier Invitrogen™ Supplier No. MA549276
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human K562 whole cell, human HepG2 whole cell, human PC-3 whole cell, human RT4 whole cell. ICC/IF: PC-3 cell. Flow: SiHa cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Probable E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. May be required for development of the head mesenchyme and neural tube closure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF5
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2F13C4
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene Eif5
Gene Accession No. P55010
Gene Alias 2810011H21Rik; D12Ertd549e; EIF5; EIF-5; EIF-5A; eukaryotic initiation factor 5 (eIF-5); eukaryotic translation initiation factor 5
Gene Symbols Eif5
Host Species Mouse
Immunogen E.coli-derived human EIF5 recombinant protein (Position: D11-D423).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1983
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.